Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium borohydride, 4M (10 wt.%) solution in THF, AcroSeal™

Catalog No. ac428881000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy

Catalog No. AC428881000

Supplier: Thermo Scientific Chemicals 428881000

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 16949-15-8 | BH4Li | 21.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lithium Borohydride
CAS 16949-15-8, 109-99-9
Quantity 100 mL
Molecular Formula BH4Li
Linear Formula LiBH4
MDL Number MFCD00011088
Flash Point −21°C
Merck Index 15,5581
Synonym borate 1-, tetrahydro-, lithium, unii-8l87x4s4kp, boryllithium, lithium, boryl, lithium hydroborate, lithotab borohydride, bh4li, lithium 1+ ion boranuide, lithium borohydride 2.0 m in thf, borate 1-, tetrahydro-, lithium 1:1
Solubility Information Solubility in water: decomposes
InChI Key UUKMSDRCXNLYOO-UHFFFAOYSA-N
SMILES [Li+].[BH4-]
IUPAC Name lithium;boron(1-)
Molecular Weight (g/mol) 21.78
PubChem CID 20722760
Formula Weight 21.77
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.89
Density 0.8900g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.