Alfa Aesar™ Lithium borohydride, 95%

Manufacturer: Alfa Aesar™ 3256309

Quantity 10g

 View more versions of this product

Catalog No. AA3256309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16949-15-8
    Linear Formula LiBH4
    Formula Weight 21.78
    MDL Number MFCD00011088
    UN Number UN1413
    Merck Index 14,5525
    Synonym Lithium tetrahydridoborate
    Chemical Name or Material Lithium borohydride

    Specifications

    Formula Weight 21.78
    Health Hazard 2 GHS H Statement
    H260-H301-H311-H331-H314-H318
    In contact with water releases flammable gases which may ignite spontaneously.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Physical Form Powder
    Linear Formula LiBH4
    Merck Index 14,5525
    Health Hazard 1 Danger
    RTECS Number ED2725000
    UN Number UN1413
    Melting Point ≈280°C (dec.)
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: LITHIUM BOROHYDRIDE
    Odor Odorless
    Sensitivity Moisture Sensitive
    MDL Number MFCD00011088
    CAS 16949-15-8
    Synonym Lithium tetrahydridoborate
    Color White
    Quantity 10g
    Solubility Information Soluble in ether, THF, and aliphatic amines
    Density 0.66g/mL
    TSCA TSCA
    Chemical Name or Material Lithium borohydride
    Assay 95%

    Description

    • Strong reducing agent
    • Used to reduce compounds containing ketones, aldehydes, ester carbonyls, acid chlorides, lactones, and epoxides

    GHS H Statement
    H260-H301-H311-H331-H314-H318
    In contact with water releases flammable gases which may ignite spontaneously.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit