Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Lithium Carbonate
SCI_ED

Catalog No. S25388 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Lab Grade
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25388 100 g Lab Grade
S25388A 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25388

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium(1+) carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Color White
Quantity 100 g
Linear Formula Li2CO3
Formula Weight 73.89
Packaging Lab Grade
Density 2.1g/mL
Chemical Name or Material Lithium Carbonate
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.