Learn More
Lithium carbonate, Puratronic™, 99.998% (metals basis excluding Ca), Ca LT 20ppm
Description
Stainless Steel powder is employed in pharmaceuticals, marine applications, architectural applications, medical implants, including pins, screws and orthopaedic implants like total hip and knee replacements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Boiling Point | 1310°C (decomposition) |
Melting Point | 723°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.998% (Metals basis excluding Ca) |
Solubility Information | Soluble in water and acid. Insoluble in acetone,ammonia and alcohol. |
IUPAC Name | dilithium;carbonate |
Formula Weight | 73.9 |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H302-H319 |
Precautionary Statement | P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c |
EINECSNumber | 209-062-5 |
RTECSNumber | OJ5800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.