Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Lithium Chloride
Shop All Fisherbrand Products

Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
Reagent Grade
Description
This product is for educational use - no C of A is available.
Specifications
Specifications
Color | White |
Physical Form | Powder, Crystals and/or Chunks |
Quantity | 100 g |
Linear Formula | LiCl |
Formula Weight | 42.39 |
Packaging | Reagent Grade |
Density | 2.07g/mL |
Chemical Name or Material | Lithium Chloride |
Safety and Handling

ShelfLife | 1 year |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction