Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Chloride, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603927 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
125 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

Catalog No. 18-603-927

Supplier: Spectrum Chemical Manufacturing Corporation L1109125GMCS6

Only null left
Add to Cart
Add to Cart

L1109, 7447-41-8, LiCl

Spectrum™ Chemical Lithium Chloride, Crystal, Reagent, ACS is a chemical compound that is extremely soluble in polar solvents and is used in order to obtain lithium metal and as a synthetic reagent. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
IUPAC Name lithium(1+) chloride
SMILES [Li+].[Cl-]

Specifications

CAS Max % 100%
Quantity 6 x 125 g
Percent Purity 99%
Loss on Drying 1%
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.