Learn More
Lithium chloride, 99%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1382.0°C |
| Melting Point | 605.0°C |
| Quantity | 1 kg |
| Assay Percent Range | 98.5% min. (Argentometry) |
| Linear Formula | LiCl |
| Fieser | 01,609; 02,246; 04,298; 05,677; 12,277; 16,194 |
| Merck Index | 15,5584 |
| Solubility Information | Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols, ether, pyridine,, nitrobenzene and acetone |
| Formula Weight | 42.38 |
| Percent Purity | 99% |
| Show More |
Frequently Asked Questions (FAQs)
Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Causes skin irritation. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse |
| EINECSNumber | 231-212-3 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.