Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium chloride hydrate, Puratronic™, 99.996% (metals basis)

Catalog No. AA1073514
Click to view available options
Quantity:
25 g
100 g

CAS: 85144-11-2 | ClLi | 42.39 g/mol

Lithium chloride is widely employed in lithium metal production. It is used as a brazing flux for aluminum in automobile parts. Its use can also be seen as a desiccant for drying air streams. In biochemical applications, lithium chloride is used to precipitate RNA from cellular extracts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85144-11-2
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00149764
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride hydrate, lithium chloride monohydrate, lithium chloride, monohydrate, lithiumchloridehydrate, hcl2lio, licl h2o, licl.h2o, acmc-1bre5, ksc182g6f, lithium chloride hydrate, puratronic
PubChem CID 23681138
SMILES [Li+].[Cl-]

Specifications

Melting Point >98°C-H2O
Color White
Physical Form Crystalline Aggregates
Quantity 25 g
Assay Percent Range 99.996% (Metals basis)
Solubility Information Soluble in water.
IUPAC Name lithium;chloride;hydrate
Formula Weight 42.39 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Lithium chloride hydrate
Show More Show Less
What storage conditions should I use for Lithium chloride?

Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.

Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 231-212-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.