Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium chloride, ultra dry, 99.995% (metals basis)

Catalog No. AA4476222
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 7447-41-8 | ClLi | 42.39 g/mol

Lithium chloride is useful for the production of lithium metal and for the generation of Mn(0) species which can be used in free radical cyclizations. It can serve as a flame colorant to generate dark red flames, a brazing flux for aluminum in automobiles, a hygrometer and a desiccant for drying air streams. On exposure to air, it becomes a solution with the concentration directly related to relative humidity of the atmosphere and hence serves as a relative humidity standard in calibrating hygrometers. Apart from being a source of chloride, it serves as an additive in the Stille reaction in organic synthesis and to precipitate RNA from cellular extracts. Being biologically significant, it finds applications in a wide variety of assays to study cell-fate and neurobiology. Lithium chloride has been found to inhibit virus infection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1325°C to 1360°C
Melting Point 605°C
Color White to Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.995% (Metal basis)
Solubility Information Soluble in water methanol,ethanol,pyridine and formic acid. Insoluble in dichloromethane.
IUPAC Name lithium;chloride
Formula Weight 42.39
Odor Odorless
Sensitivity Hygroscopic
Appearance White to gray crystalline powder
Density 2.068 g/cm3 at 20°C
Chemical Name or Material Lithium chloride, ultra dry
Show More Show Less
What storage conditions should I use for Lithium chloride?

Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.

Hazard Category H302-H315-H319
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
EINECSNumber 231-212-3
RTECSNumber OJ5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.