Learn More
Lithium cyclopentadienide, 97%
Description
Lithium cyclopentadienide is a useful reagent in preparing metallocenes and cyclopentadienyl derivatives. It is also used as intermediates and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 5 g |
UN Number | UN3180 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 72.04 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | Lithium cyclopentadienide |
Safety and Handling
Hazard Category | H314 |
Hazard Statement | GHS H Statement H228-H314-H318 Flammable solid. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: II |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.