Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Lithium Diisopropylamide Solution
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
4 x 100 mL
Description
Lithium diisopropylamide solution (LDA) is a sterically hindered non-nucleophilic strong base widely used in organic synthesis.Lithium diisopropylamide solution (LDA) can be used:As an initiator in the anionic polymerization of D,L-lactide and methyl methacrylate.To facilitate ester enolization.To convert carboxylic acids to enediolate intermediates for preparing trifluoromethyl ketones.In ortho-lithiation of arylsulfonyloxazolidinones to prepare N-substituted saccharin analogs.To catalyze ortholithiation and Fries rearrangement of aryl carbamates.As a promoter in the isomerization of allylic ethers to (Z)-propenyl ethers.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
Specifications
Specifications
| CAS | 4111-54-0 |
| Density | 0.812 g/mL at 25°C |
| Molecular Formula | C6H14LiN |
| Linear Formula | [(CH3)2CH]2NLi |
| MDL Number | MFCD00064449 |
| Quantity | 4 x 25 mL |
| Synonym | LDA |
| Molecular Weight (g/mol) | 107.12 g/mol |
| Biological Activity | Hearing Organs |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction