Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium dimethylamide, 96%

Catalog No. p-7059186
Click to view available options
Quantity:
5 g
25 g

CAS: 3585-33-9 | C2H6LiN | 51.017 g/mol

It is used in the synthesis of some d0 organoimido complexes of the early transition metals and in meta rearrangement in the reaction of triphenyl-(p-bromophenyl)-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3585-33-9
Molecular Formula C2H6LiN
Molecular Weight (g/mol) 51.017
MDL Number MFCD00008295
InChI Key YDGSUPBDGKOGQT-UHFFFAOYSA-N
Synonym lithium dimethylamide, lithium dimethylazanide, linme2, acmc-20akms, dimethylamino lithium, lithiodimethylamine, lithium dimethylamide preparation, lithium 1+ dimethylazanide, lithium dimethylamide w/v in hexane
PubChem CID 5152226
IUPAC Name lithium;dimethylazanide
SMILES [Li+].C[N-]C

Specifications

Melting Point (decomposition)
Color White
Odor Amine-like
Linear Formula LiN(CH3)2
Quantity 5 g
UN Number UN3393
Beilstein 3618233
Sensitivity Air and moisture sensitive
Solubility Information Reacts violently with water.
Formula Weight 51.02
Percent Purity 96%
Physical Form Powder or Lumps
Chemical Name or Material Lithium dimethylamide
Show More Show Less
Hazard Category H250-H261-H314-H335
Hazard Statement GHS H Statement
H250-H261-H314-H318
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P222-P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC, WATER-REACTIVE
EINECSNumber 222-714-3
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.