Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium fluoride

Catalog No. 4499032G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
2 g
10 g
Catalog No. Quantity
44-990-32G 2 g
44-990-310G 10 g
2 options

Catalog No. 44-990-32G

Supplier: Sigma Aldrich 4499032G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium fluoride

Specifications

CAS 7789-24-4
Melting Point 845°C
Density 2.64 g/mL (at 25°C)
Boiling Point 1673°C
Molecular Formula FLi
Linear Formula LiF
Quantity 2 g
MDL Number MFCD00011090
Synonym Fluorolithium
Molecular Weight (g/mol) 25.94
Percent Purity ≥99.99% trace metals basis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.