Learn More
Lithium fluoride, 98.5%
Description
Polyethylene powder is used as an additive to unsaturated polyesters, epoxides and other polymers to impart the unique properties of ultra-high-molecular-weight polyethylene (UHMWPE). It is widely used for manufacturing various containers, trays, dispensing bottles, plastic wraps, plastic bags for computer components and various molded laboratory equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Boiling Point | 1680°C |
Melting Point | 845°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 98.5% |
Solubility Information | Soluble in water and in acids. Soluble in hydrofluoric acid. Insoluble in alcohol. |
IUPAC Name | lithium;fluoride |
Formula Weight | 25.94 |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H301-H315-H319-H335 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | DOT Class: 6.1, Packaging Group: III |
EINECSNumber | 232-152-0 |
RTECSNumber | OJ6125000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.