Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium fluoride, 99.85% (metals basis)

Catalog No. p-7050270
Click to view available options
Quantity:
10 g
50 g

CAS: 7789-24-4 | FLi | 25.94 g/mol

Used in the preparation of hypersensitive thermoluminescent materials, dosimeters and in the room-temperature oxidation of noble metals using fluorine in anhydrous HF. It is mainly used as a component of molten salts. Lithium fluoride forms the basic constituent of the preferred fluoride salt mixture used in liquid-fluoride nuclear reactors. Lithium fluoride is widely used in PLED and OLED as a coupling layer to enhance electron injection. LiF crystals are transparent to short wavelength ultraviolet radiation, more so than any other material. LiF is therefore used in specialized UV optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-24-4
Molecular Formula FLi
Molecular Weight (g/mol) 25.94
MDL Number MFCD00011090
InChI Key PQXKHYXIUOZZFA-UHFFFAOYSA-M
Synonym lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PubChem CID 224478
IUPAC Name lithium;fluoride
SMILES [Li+].[F-]

Specifications

Boiling Point 1680°C
Melting Point 845°C
Color White
Physical Form Lump/Powder
Quantity 10 g
Linear Formula LiF
UN Number UN3288
Merck Index 14,5531
Solubility Information Soluble in 0.29g/100 mL (20°C) and hydrogen fluoride. Insoluble in alcohol.
Formula Weight 25.94
Percent Purity 99.85%
Odor Odorless
Refractive Index 1.3915
Density 2.635 g/mL
Assay (metals basis)
Chemical Name or Material Lithium fluoride
Show More Show Less
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 232-152-0
RTECSNumber OJ6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.