Alfa Aesar™ Lithium fluoride, Optical Grade, 99.5+%

Manufacturer: Alfa Aesar™ 1405606

Quantity 5g

 View more versions of this product

Catalog No. AA1405606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-24-4
    Formula Weight 25.94
    MDL Number MFCD00011090
    UN Number UN3288
    Merck Index 14,5531
    Chemical Name or Material Lithium fluoride
    Grade Optical Grade

    Specifications

    Formula Weight 25.94
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    UN Number UN3288
    Melting Point 845°C
    Quantity 5g
    Physical Form Broken Crystals
    MDL Number MFCD00011090
    Merck Index 14,5531
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    CAS 7789-24-4
    Boiling Point 1680°C
    TSCA TSCA
    Refractive Index 1.3915
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 2.635g/mL
    Chemical Name or Material Lithium fluoride
    Assay ≥99.5%
    Grade Optical Grade

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit