Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium hexafluoroantimonate, 97%, Thermo Scientific Chemicals

Catalog No. p-7050551
Click to view available options
Quantity:
25 g

Lithium hexafluoroantimonate as a photo initiator for production of polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18424-17-4
Molecular Formula F6LiSb
Molecular Weight (g/mol) 242.69
MDL Number MFCD00042319
InChI Key YVBBFYDVCPCHHG-UHFFFAOYSA-H
Synonym lithium hexafluoroantimonate, lithium 1+ ion hexafluorostibanuide, acmc-1bpt7, lithotab hexafluorostibanuide, lithium hexafluorostiboranuide, hexafluorostibine, lithium salt, lithium hexafluoroantimonate 1-, lithium hexakis fluoranyl antimony 1-
PubChem CID 16700586
IUPAC Name lithium;hexafluoroantimony(1-)
SMILES [Li+].F[Sb-](F)(F)(F)(F)F

Specifications

Boiling Point (decomposition)
Physical Form Powder
Quantity 25 g
UN Number UN1549
Solubility Information Very soluble in water
Formula Weight 242.68
Percent Purity 97%
Sensitivity Hygroscopic
Chemical Name or Material Lithium hexafluoroantimonate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.