Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium hydride, 97%, pure, powder (<100 μm), Thermo Scientific™

Catalog No. AC191191000
Click to view available options
Quantity:
10 g
100 g
Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7580-67-8
Molecular Formula HLi
Molecular Weight (g/mol) 7.95
MDL Number MFCD00011074
InChI Key SRTHRWZAMDZJOS-UHFFFAOYSA-N
Synonym lithium hydride, lithiumhydride, lih, lithium hydride anion, hydride lithium, hydridolithate 1-, lih-, ksc491a4n
PubChem CID 62714
IUPAC Name lithium;hydride
SMILES [H-].[Li+]

Specifications

Melting Point 680.0°C
Color White to Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 96% min
Linear Formula LiH
Fieser 04,304; 13,165
Merck Index 15, 5589
Solubility Information Solubility in water: reacts. Other solubilities: insoluble in benzene and toluene, soluble in ether
Formula Weight 7.95
Percent Purity 97%
Grade Pure
Specific Gravity 0.82
Density 0.8200g/mL
Chemical Name or Material Lithium hydride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Handle under inert gas.
Protec
EINECSNumber 231-484-3
RTECSNumber OJ6300000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.