Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium hydride, 99.4% (metals basis)

Catalog No. p-7051584
Click to view available options
Quantity:
25 g
100 g

CAS: 7580-67-8 | HLi | 7.948 g/mol

Lithium hydride is used as a precursor in the production of a variety of reagents in the synthesis, especially silane by reacting with silicon tetrachloride through Sundermeyer process and lithium aluminum hydride and lithium borohydride. It finds application in nuclear reactors as a shielding material. In addition, it is a hydrogen source or reducing agent to prepare other hydrides amides and 2H isotopic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7580-67-8
Molecular Formula HLi
Molecular Weight (g/mol) 7.948
MDL Number MFCD00011074
InChI Key SRTHRWZAMDZJOS-UHFFFAOYSA-N
Synonym lithium hydride, lithiumhydride, lih, lithium hydride anion, hydride lithium, hydridolithate 1-, lih-, ksc491a4n
PubChem CID 62714
IUPAC Name lithium;hydride
SMILES [H-].[Li+]

Specifications

Melting Point 686°C
Color White to Gray
Physical Form Powder
Quantity 25 g
Linear Formula LiH
UN Number UN1414
Merck Index 14,5533
Solubility Information Slightly soluble in dimethylformamide. Insoluble in acetone,benzene and toluene.
Formula Weight 7.95
Percent Purity 99.4%
Odor Odorless
Sensitivity Air and moisture sensitive
Density 0.78 g/mL
Assay (metals basis)
Chemical Name or Material Lithium hydride
Show More Show Less
Hazard Category H261-H314
Hazard Statement GHS H Statement
H260-H301-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: LITHIUM HYDRIDE
EINECSNumber 231-484-3
RTECSNumber OJ6300000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.