Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium hydroxide, 98%, pure, anhydrous, Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
1310-65-2
LiOH
23.95
MFCD00011095
WMFOQBRAJBCJND-UHFFFAOYSA-M
lithium hydroxide,lithium hydrate,lioh,lithium hydroxide anhydrous,lithium hydroxide li oh,lithiumhydroxid,lithiumhydroxide,lithium hydoxide,unii-903yl31jas,lithium hydroxide, anhydrous
3939
CHEBI:33979
lithium;hydroxide
[Li+].[OH-]
1310-65-2
LiOH
23.95
MFCD00011095
WMFOQBRAJBCJND-UHFFFAOYSA-M
lithium hydroxide,lithium hydrate,lioh,lithium hydroxide anhydrous,lithium hydroxide li oh,lithiumhydroxid,lithiumhydroxide,lithium hydoxide,unii-903yl31jas,lithium hydroxide, anhydrous
3939
CHEBI:33979
lithium;hydroxide
[Li+].[OH-]
Assay Percent Range 98%
Merck Index 15,559
Solubility Information Solubility in water: 113g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 23.95
Physical Form Crystalline Powder
Percent Purity 98%
Grade Pure
Density 1.4300g/mL
Packaging Plastic bottle
Solubility Solubility in water: 113g/L (20°C). Other solubilities: slightly soluble in alcohol
Specific Gravity 1.43
Color White to Yellow
Melting Point 462.0°C
Quantity 100g
Chemical Name or Material Lithium hydroxide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 215-183-4