Learn More
Lithium hydroxide, anhydrous, 98%
Description
Ammonium peroxydisulfate is widely utilized as an oxidizing agent and radical initiators in the polymerization of certain alkenes. It is actively involved in in the synthesis of commercially important polymers like styrene-butadiene rubber and polytetrafluoroethylene. It combines with tetramethylethylenediamine to catalyze the polymerization of acrylamide. Due to its oxidizing properties, it can be employed to etch copper on printed circuit boards which is an alternative to ferric chloride solution. Also, it acts as oxidants in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Boiling Point | 924°C |
Melting Point | 462°C |
Color | White |
Quantity | 100 g |
Assay Percent Range | 98% |
Solubility Information | Soluble in water. Slightly soluble in alcohol. |
IUPAC Name | lithium;hydroxide |
Formula Weight | 23.95 |
Odor | Odorless |
Sensitivity | Air sensitive; Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H302-H314-H335-H336-H373 |
Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | DOT Class: 8, Packaging Group: II |
EINECSNumber | 215-183-4 |
RTECSNumber | OJ6307070 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.