Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium hydroxide monohydrate

Catalog No. 45019725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
25 g

Catalog No. 45-019-725G

Supplier: Sigma Aldrich 45019725G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium hydroxide monohydrate (LiOH.H2O) is a lithium source that can be prepared from lithium carbonate by membrane electrolysis.LiOH.H2O can be used in the preparation of porous cathode films for the fabrication of lithium batteries. It can also be used in the development of lithium oxygen batteries.

Specifications

CAS 1310-66-3
Molecular Formula HLiO -+ H2O
Linear Formula LiOH -+ H2O
MDL Number MFCD00149772
Quantity 25 g
Synonym Lithium hydroxide hydrate
Molecular Weight (g/mol) 41.96
Percent Purity 99.995% trace metals basis
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.