Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium hydroxide monohydrate, battery grade

Catalog No. p-7032983
Click to view available options
Quantity:
250 g
1000 g

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol

Lithium hydroxide monohydrate is used in preparation of other lithium salts, as a catalyst in the production of alkyd resins, in esterifications. It is also used in the production of lithium soaps, greases, sulfonates and electric storage batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium hydroxide monohydrate is used in preparation of other lithium salts, as a catalyst in the production of alkyd resins, in esterifications. It is also used in the production of lithium soaps, greases, sulfonates and electric storage batteries.

Solubility
Soluble in water (216 mg/ml at 20°C), ethanol (freely), and alcohol (slightly).

Notes
Air sensitive and hygroscopic. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
IUPAC Name lithium;hydroxide;hydrate
SMILES [Li+].O.[OH-]

Specifications

Quantity 250 g
UN Number UN2680
Solubility Information Soluble in water (216mg/ml at 20°C),ethanol (freely),and alcohol (slightly).
Formula Weight 41.96(23.95 Anhydrous)
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material Lithium hydroxide monohydrate
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H331-H314-H318
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: LITHIUM HYDROXIDE
EINECSNumber 215-183-4
RTECSNumber OJ6307070
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.