Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Hydroxide Monohydrate, Solstice

Catalog No. 60044772 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 1 kg
100 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-772 6 x 1 kg
60-009-05 100 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-772 Supplier Solstice Supplier No. 130206X1KG

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Purum,≥98.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
SMILES [Li+].O.[OH-]

Specifications

Quantity 6 x 1 kg
Linear Formula LiOH · H2O
UN Number UN2680
IUPAC Name lithium(1+) hydrate hydroxide
Formula Weight 41.96
Percent Purity ≥98.5%
Grade purum
Chemical Name or Material Lithium Hydroxide Monohydrate
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H302-H314
DOTInformation 8 / PGII
EINECSNumber 215-183-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.