Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium iodide, 99%, pure, anhydrous

Catalog No. p-222308
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle

CAS: 10377-51-2 | ILi | 133.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-51-2
Molecular Formula ILi
Molecular Weight (g/mol) 133.84
MDL Number MFCD00011092
InChI Key HSZCZNFXUDYRKD-UHFFFAOYSA-M
Synonym lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
PubChem CID 66321
IUPAC Name lithium;iodide
SMILES [Li+].[I-]

Specifications

Boiling Point 1171.0°C
Melting Point 446.0°C
Physical Form Powder, Crystals and/or Granules
Quantity 50 g
Assay Percent Range 98.5% min. (on anhydrous substance) (Argentometry)
Linear Formula LiI
Fieser 01,615; 04,304; 05,410; 07,208; 09,283; 10,245; 11,300; 12,282; 15,185; 16,197; 17,85
Merck Index 15, 5591
Solubility Information Solubility in water: 165 g/100 mL (20°C). Other solubilities: soluble in alcohol, freely soluble in acetone
Formula Weight 133.85
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Density 3.49
Chemical Name or Material Lithium iodide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at
EINECSNumber 233-822-5
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.