Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ Lithium Ion Battery Storage Application Label for Hazardous Material Safety Cabinets
Shop All Justrite Safety Group Products
Click to view available options
For Use With (Equipment):
Hazardous Material Cabinet
Specifications
Specifications
| For Use With (Equipment) | Hazardous Material Cabinet |
| Product Type | Lithium-ion Battery Storage Application Label |
| Height (English) Exterior | 12 in. |
| Color | Black |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction