Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium manganese(III,IV) oxide, 99.5% (metals basis)

Catalog No. p-7051011
Click to view available options
Quantity:
25 g
100 g

CAS: 12057-17-9 | LiMn2O4 | 180.81 g/mol

It is used as anode materials of high-performance lithium ion battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12057-17-9
Molecular Formula LiMn2O4
Molecular Weight (g/mol) 180.81
MDL Number MFCD01114233
InChI Key KEZPOJYZIWMPGY-UHFFFAOYSA-N

Specifications

Melting Point >400°C
Physical Form Powder
Quantity 25 g
Linear Formula LiMn2O4
Solubility Information Insoluble in water.
Formula Weight 180.81
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Lithium manganese(III,IV) oxide
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.