Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium metaborate, anhydrous, 99.9% (metals basis)

Catalog No. p-4407577
Click to view available options
Quantity:
50 g
250 g
1 kg

CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol

Used in borate fusion sample preparation of various samples for analysis by XRF, AAS, ICP-MS, ICP-AES and ICP-OES. Used to determine the trace level of elements in contaminated soil. Useful fod dissolving acidic oxides such as SiO2, and Fe2O3. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-69-5
Molecular Formula B4H14Li2O7
Molecular Weight (g/mol) 183.23
MDL Number MFCD00011089
InChI Key DPYKRXYVXDYLEY-UHFFFAOYSA-N
Synonym lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
PubChem CID 123308
SMILES [Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O

Specifications

Melting Point 845°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in ethanol. Partially soluble in cold water.
IUPAC Name tetraboron heptahydrate dilithium
Formula Weight 49.75
Sensitivity Hygroscopic
Density 1.397
Chemical Name or Material Lithium metaborate, anhydrous
EINECSNumber 236-631-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.