Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium methoxide, pure, 2.2M (10 wt%) solution in methanol, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
AcroSeal™ Glass Bottle in Tin Can
865-34-9,67-56-1
CH3LiO
37.974
MFCD00036357
JILPJDVXYVTZDQ-UHFFFAOYSA-N
lithium methoxide,lithium methanolate,methanol, lithium salt,lithium methylate,liome,methanol, lithium salt 1:1,liom,lithium methylat,lithotab methoxide,lioch3
4177230
lithium;methanolate
[Li+].C[O-]
865-34-9,67-56-1
CH3LiO
37.974
MFCD00036357
JILPJDVXYVTZDQ-UHFFFAOYSA-N
lithium methoxide,lithium methanolate,methanol, lithium salt,lithium methylate,liome,methanol, lithium salt 1:1,liom,lithium methylat,lithotab methoxide,lioch3
4177230
lithium;methanolate
[Li+].C[O-]
Linear Formula CH3OLi
Density 0.8500g/mL
Formula Weight 37.97
Physical Form Liquid
Percent Purity ≥10%
Grade Pure
Packaging AcroSeal™ Glass Bottle
Color Colorless
Quantity 800mL
Chemical Name or Material Lithium methoxide, 10% Solution in Methanol

GHS P Statement
Causes damage to organs.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic in contact with skin.
Highly flammable liquid and vapor.
Self-heating; may catch fire.
Reacts violently with water.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF exposed: Call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 212-737-7

TSCA : TSCA