Learn More
Lithium niobium ethoxide, 99+% (metals basis), 5% w/v in ethanol, Thermo Scientific Chemicals
Description
Lithium niobium ethoxide is used in the preparation of lithium niobate, which is a ferroelectric material used as an optical switch. It is also used as solid electrolytes due to its high ionic diffusivity in the field of material science.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Flash Point | 17°C (63°F) |
Odor | Alcohol-like |
Quantity | 25 mL |
UN Number | UN1170 |
Sensitivity | Moisture sensitive |
Formula Weight | 370.21 |
Percent Purity | ≥99% |
Physical Form | Liquid |
Assay | (metals basis) |
Show More |
Safety and Handling
Hazard Category | H225-H314-H335 |
Hazard Statement | GHS H Statement H225-H315-H319 Highly flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
For Manufacturing and Laboratory Use. Not for beverage use
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.