Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Lithium Nitrate
SCI_ED

Catalog No. S25392 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25392 100 g Reagent Grade
S25392A 500 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25392

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
PubChem CID 10129889
ChEBI CHEBI:63315
IUPAC Name lithium(1+) nitrate
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Color White
Quantity 100 g
Assay Percent Range >95%
Linear Formula LiNO3
Formula Weight 68.95
Grade Reagent
Packaging Reagent Grade
Density 2.38g/mL
Chemical Name or Material Lithium Nitrate
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 5.1, III, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.