Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium nitrate, anhydrous, 99.999% (metals basis)

Catalog No. AA4445609
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 7790-69-4 | LiNO3 | 68.94 g/mol

Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent used in the production of red-colored fireworks and flares, as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from sun for cooking. It is applied to concrete-pavement to withstand weathering effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
PubChem CID 10129889
ChEBI CHEBI:63315
IUPAC Name lithium;nitrate
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Boiling Point 600°C (decomposition)
Melting Point 264°C
Color White
Physical Form Crystalline/Lump
Quantity 10 g
UN Number UN2722
Merck Index 14,5536
Solubility Information Soluble in water,ethanol,methanol,pyridine,ammonia and acetone.
Formula Weight 68.95
Percent Purity 99.999%
Odor Odorless
Density 2.38 g/mL
Assay (metals basis)
Chemical Name or Material Lithium nitrate, Anhydrous
Show More Show Less
Hazard Category H272-H302-H319
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: LITHIUM NITRATE
EINECSNumber 232-218-9
RTECSNumber QU9330000
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.