Lithium nitrate hydrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1074209

Quantity 10g

 View more versions of this product

Catalog No. AA1074209

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Crystalline Aggregates
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula LiNO3·xH2O (x≈0.5)
    MDL Number MFCD00011094
    Chemical Name or Material Lithium nitrate hydrate

    Specifications

    Quantity 10g
    EINECS Number 232-218-9
    Physical Form Crystalline Aggregates
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula LiNO3·xH2O (x≈0.5)
    MDL Number MFCD00011094
    Chemical Name or Material Lithium nitrate hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit