Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium peroxide, 95%

Catalog No. p-7049227
Click to view available options
Quantity:
100 g
500 g

CAS: 12031-80-0 | Li2O2 | 45.878 g/mol

Lithium peroxide is used in air purifiers. It is also used to produce high-purity oxygen. It acts as a catalyst for polymerization of styrene to polystyrene. Further, it serves as a curing agent for special polymers and a source of oxygen in sealed spaces such as submarines and in breathing apparatus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-80-0
Molecular Formula Li2O2
MDL Number MFCD00016184
InChI Key HPGPEWYJWRWDTP-UHFFFAOYSA-N
PubChem CID 25489
IUPAC Name dilithium;peroxide
SMILES [Li+].[Li+].[O-][O-]

Specifications

Color White
Physical Form Powder
Quantity 100 g
UN Number UN1472
Synonym dilithium peroxide, lithium peroxide li2 o2, lithium oxide, unii-9anx556r5f, lithium peroxide un1472 oxidizer
Solubility Information Soluble in anhydrous acetic acid. Insoluble in alcohol.
Molecular Weight (g/mol) 45.878
Formula Weight 45.88
Percent Purity 95%
Odor Odorless
Sensitivity Air and moisture sensitive
Chemical Name or Material Lithium peroxide
Show More Show Less
Hazard Category H272-H314
Hazard Statement GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: LITHIUM PEROXIDE
EINECSNumber 234-758-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.