Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium phosphate, extra pure

Catalog No. AC389785000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

CAS: 10377-52-3 | Li3O4P | 115.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-52-3
Molecular Formula Li3O4P
Molecular Weight (g/mol) 115.79
MDL Number MFCD00016187
InChI Key TWQULNDIKKJZPH-UHFFFAOYSA-K
Synonym lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4
PubChem CID 165867
SMILES [Li+].[Li+].[Li+].[O-]P([O-])([O-])=O

Specifications

Melting Point 837°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Linear Formula Li3PO4
Solubility Information Solubility in water: 0.4g/L (20°C)
IUPAC Name trilithium;phosphate
Formula Weight 115.79
Percent Purity ≥95.0%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Lithium phosphate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-823-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.