Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lithium Standard, 3000 mEq/L, Reagents
Click to view available options
Quantity:
4 L
20 L
Specifications
Specifications
Chemical Name or Material | Lithium Standard, 3000 mEq/L |
Packaging | Glass Bottle |
CAS | 7732-18-5, 7447-41-8 |
Specific Gravity | 1.07 |
Physical Form | Liquid |
Boiling Point | 103°C |
Solubility | Completely miscible |
Quantity | 4 L |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction