Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Plastic bottle
7439-93-2,7647-01-0,7732-18-5
Li
6.94
MFCD00134051
WHXSMMKQMYFTQS-UHFFFAOYSA-N
3028194
CHEBI:30145
lithium
[Li]
7439-93-2,7647-01-0,7732-18-5
Li
6.94
MFCD00134051
WHXSMMKQMYFTQS-UHFFFAOYSA-N
3028194
CHEBI:30145
lithium
[Li]
For atomic adsorption spectroscopy
Concentration 995 to 1005 μg/mL of Li (1+) in 2% HCl (20°C)
Formula Weight 6.94
Physical Form Solution
Grade AAS
Packaging Plastic bottle
Density 0.53
Color Undesignated
Quantity 100mL
Chemical Name or Material Lithium standard solution, 1 mg/ml Li in 2% HCl

GHS H Statement:
May be corrosive to metals.

GHS P Statement:
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning