Learn More
Lithium sulfate, anhydrous, 99.7% (metals basis)
Description
Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | 845°C |
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Assay Percent Range | 99.7% (Metal basis) |
Solubility Information | Soluble in water. Insoluble in acetone,ethanol and pyridine. |
IUPAC Name | dilithium(1+) sulfate |
Formula Weight | 109.94 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H302-H319 |
Precautionary Statement | P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c |
EINECSNumber | 233-820-4 |
RTECSNumber | OJ6419000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.