Learn More
Lithium sulfate, anhydrous, 99.7% (metals basis), Thermo Scientific Chemicals
Description
Lithium sulfate is used in the treatment of bipolar disorder. It acts as an important component of ion conducting glasses, solar panels and in battery. It serves as an additive for Portland cement. Further, it is used in the production of special high-strength glass, food and flavoring agent in synthetic beverages. In addition to this, it is used as a catalyst in organic synthesis for the preparation of 1-butene from n-butyl bromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 845°C |
Color | White |
Physical Form | Powder |
Quantity | 5 kg |
Assay Percent Range | 99.7% (Metal basis) |
Solubility Information | Soluble in water. Insoluble in acetone,ethanol and pyridine. |
IUPAC Name | dilithium;sulfate |
Formula Weight | 109.94 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | H302-H319 |
missing translation for 'healthHazard3' | P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c |
EINECSNumber | 233-820-4 |
RTECSNumber | OJ6419000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.