Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium sulfate, anhydrous, 99.99% (metals basis)

Catalog No. p-7059296
Click to view available options
Quantity:
25 g
100 g

CAS: 10377-48-7 | Li2O4S | 109.94 g/mol

Lithium sulfate, anhydrous is used to treat bipolar disorder, solar panels and the potential for a new class of battery, and used as a catalyst for the elimination reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-48-7
Molecular Formula Li2O4S
Molecular Weight (g/mol) 109.94
MDL Number MFCD00011086
InChI Key INHCSSUBVCNVSK-UHFFFAOYSA-L
Synonym lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
PubChem CID 66320
ChEBI CHEBI:53474
SMILES [Li+].[Li+].[O-]S([O-])(=O)=O

Specifications

Melting Point 845°C
Color White
Physical Form Coarse Powder
Quantity 25 g
Merck Index 14,5541
Solubility Information Soluble in water(345g/L). Insoluble in absolute ethanol,acetone and pyridine.
IUPAC Name dilithium(1+) sulfate
Formula Weight 109.94
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.22 g/mL
Assay (metals basis)
Chemical Name or Material Lithium sulfate, Anhydrous
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 233-820-4
RTECSNumber OJ6419000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.