Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium sulfate monohydrate, ACS, 99.0% min

Catalog No. AA3621636
Click to view available options
Quantity:
100 g
500 g
2 kg

Maximum level of impurities: Loss on drying at 150°C 13.0-15.0%, Insoluble matter 0.01%, Cl 0.002%, NO3 0.001%, Heavy Metals (as Pb) 0.001%, Fe 0.001%, K 0.05%, Na 0.05% | CAS: 10102-25-7 | H2Li2O5S | 127.95 g/mol

Lithium sulfate monohydrate is used in solar panels and potential for a new class of battery. It is also used to treat bipolar disorder and in ultrasound-type non-destructive testing. It is also used as an additive for portland cement to speed up the curing. It is also involved as a catalyst in the elimination reaction for the conversion of n-butyl bromide to 1-butene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-25-7
Molecular Formula H2Li2O5S
Molecular Weight (g/mol) 127.95
MDL Number MFCD00149766
InChI Key RKGLUDFWIKNKMX-UHFFFAOYSA-L
Synonym lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
PubChem CID 165815
SMILES [Li+].[Li+].O.[O-]S([O-])(=O)=O

Specifications

Melting Point 130°C-H2O
Color Colorless
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.0% min.
Solubility Information Soluble in water. Soluble in water. Insoluble in ethanol,acetone and pyridine.
IUPAC Name dilithium(1+) hydrate sulfate
Formula Weight 127.96 (109.94 Anhydrous)
Grade ACS
Odor Odorless
Appearance Colorless granules
Density 2.06 g/cm3 at 20°C
Chemical Name or Material Lithium sulfate monohydrate
Show More Show Less
Hazard Category H302-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 233-820-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.