Lithium sulfide, 99.9% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1283904

Quantity 2g

 View more versions of this product

Catalog No. AA1283904

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12136-58-2
    Physical Form Powder
    Assay Percent Range 99.9% (Metal basis)
    Molecular Formula Li2S
    Formula Weight 45.95
    MDL Number MFCD00011085
    Chemical Name or Material Lithium sulfide

    Specifications

    Formula Weight 45.95
    Quantity 2g
    Color Yellow
    DOT Information DOT Class: 4.3, Packaging Group: II
    Density 1.66g/cm3 at 20°C
    EINECS Number 235-228-1
    Physical Form Powder
    Assay Percent Range 99.9% (Metal basis)
    Molecular Formula Li2S
    MDL Number MFCD00011085
    Health Hazard 1 H228-H261-H315-H319-H335: Flammable solid. In contact with water releases flammable gas. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Odor Like rotten eggs
    Appearance Yellow powder
    CAS 12136-58-2
    Chemical Name or Material Lithium sulfide

    H228-H261-H315-H319-H335: Flammable solid. In contact with water releases flammable gas. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit