Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tantalum oxide, 99.9% (metals basis)

Catalog No. AA4183409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4183409 10 g
AA4183418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4183409 Supplier Thermo Scientific Chemicals Supplier No. 04183409
Add to Cart

CAS: 12031-66-2 | LiO3Ta | 235.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-66-2
Molecular Formula LiO3Ta
Molecular Weight (g/mol) 235.88
MDL Number MFCD00016174
InChI Key JNQQEOHHHGGZCY-UHFFFAOYSA-N
Synonym lithium tantalate, lithium oxido dioxo tantalum, lithium 1+ tantalumoylolate
PubChem CID 5148101
IUPAC Name lithium;oxido(dioxo)tantalum
SMILES [Li+].[O--].[O--].[O--].[Ta+5]

Specifications

Physical Form Powder
Quantity 10 g
Formula Weight 235.89
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Lithium tantalum oxide
EINECSNumber 234-757-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.