Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tert-butoxide, 2.2M solution in THF, stabilized, AcroSeal™

Catalog No. p-3212811
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle

CAS: 1907-33-1 | C4H9LiO | 80.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1907-33-1, 109-99-9
Molecular Formula C4H9LiO
Molecular Weight (g/mol) 80.06
MDL Number MFCD00050479
InChI Key LZWQNOHZMQIFBX-UHFFFAOYSA-N
Synonym lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
PubChem CID 23664764
IUPAC Name lithium;2-methylpropan-2-olate
SMILES [Li+].CC(C)(C)[O-]

Specifications

Color Yellow to Brown
Density 0.8900g/mL
Flash Point −22°C
Packaging AcroSeal™ Glass bottle
Linear Formula LiOCH(CH3)3
Quantity 100 mL
Specific Gravity 0.89
Solubility Information Solubility in water: vigorous reaction
Formula Weight 80.06
Physical Form Solution
Chemical Name or Material Lithium tert-butoxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
May cause respiratory irritation.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 217-611-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.