Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lithium tetrachlorocuprate, 0.1M in THF

Manufacturer:  Alfa Aesar™ 20620AE

Quantity 100mL

 View more versions of this product

Catalog No. AA20620AE


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

15489-27-7
MFCD00011081
Lithium tetrachlorocuprate
219.24
UN2056

Specifications

15489-27-7
219.24
UN2056
0.910g/mL
Dark orange
Lithium tetrachlorocuprate
Liquid
MFCD00011081
0.1M in THF
Moisture Sensitive
100mL

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H318-H351-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit