Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tetrafluoroborate, 98%, pure

Catalog No. p-44365
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle

CAS: 14283-07-9 | BF4Li | 93.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14283-07-9
Molecular Formula BF4Li
Molecular Weight (g/mol) 93.74
MDL Number MFCD00011087
InChI Key UFXJWFBILHTTET-UHFFFAOYSA-N
Synonym lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n
PubChem CID 4298216
IUPAC Name lithium(1+) tetrafluoroboranuide
SMILES [Li+].F[B-](F)(F)F

Specifications

Melting Point 275°C
Color Gray to White
Physical Form Crystalline Powder and/or Crystals
Quantity 10 g
Assay Percent Range 98%
Linear Formula LiBF4
Fieser 05,416; 10,248
Merck Index 15, 5599
Solubility Information Solubility in water: soluble
Formula Weight 93.75
Percent Purity 98%
Grade Pure
Packaging Plastic bottle
Specific Gravity 0.852
Density 0.8520g/mL
Chemical Name or Material Lithium tetrafluoroborate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-178-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.