Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium triethylborohydride, 1M in THF, Thermo Scientific™

Catalog No. p-7080497
Click to view available options
Quantity:
500 mL

Catalog No. AA89144AP

Supplier: Thermo Scientific Chemicals 89144AP

As reducing agent

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22560-16-3
Molecular Formula C6H16BLi
Molecular Weight (g/mol) 105.94
MDL Number MFCD00011703
InChI Key WCJAYABJWDIZAJ-UHFFFAOYSA-N
Synonym Lithium triethylhydridoborate
IUPAC Name lithium(1+) triethylboranuide
SMILES [Li+].CC[BH-](CC)CC

Specifications

Color Colorless
Density 0.892 g/mL
Flash Point −17°C (1°F)
Linear Formula LiB(CH2CH3)3H
Quantity 500 mL
UN Number UN3399
Beilstein 4151240
Sensitivity Moisture sensitive
Merck Index 14,5544
Solubility Information Miscible with tetrahydrofuran and benzene.
Formula Weight 105.94
Concentration or Composition (by Analyte or Components) 1M in THF
Physical Form Liquid
Chemical Name or Material Lithium triethylborohydride
Show More Show Less
Hazard Category H225-H250-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H260-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.