Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIV-1/Zip6 Antibody (13F1), Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP326316100UL
Description
LIV-1/Zip6 Monoclonal antibody specifically detects LIV-1/Zip6 in Human samples. It is validated for ELISASpecifications
LIV-1/Zip6 | |
Monoclonal | |
Unconjugated | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
Human | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
13F1 | |
ELISA | |
zinc transporter ZIP6, LIV-1, solute carrier family 39 (zinc transporter), member 6 | |
Recombinant Human LIV-1/Zip6 protein [UniProt Q13433] | |
100 μL | |
Cancer, Endocrinology, Signal Transduction | |
25800 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction