Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIV-1/Zip6 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297843MFV500
Description
LIV-1/Zip6 Polyclonal antibody specifically detects LIV-1/Zip6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
LIV-1/Zip6 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
zinc transporter ZIP6, LIV-1, solute carrier family 39 (zinc transporter), member 6 | |
Produced in rabbits immunized with E. coli-derived Human LIV-1/Zip6 fragment. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
25800 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction