Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ LIVE/DEAD™ Fixable Violet Dead Cell Stain Kit, for 405 nm excitation

Catalog No. p-7076038
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 Assays
400 Assays
80 Assays
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L34955 200 Assays
L34963 80 Assays
50-113-8086 400 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L34955 Supplier Molecular Probes™ Supplier No. L34955
Only null left
Add to Cart
Edge
Add to Cart

The LIVE/DEAD™ Fixable Violet Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.

  • Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
  • Kit has been optimized and validated for use with a violet laser flow cytometer
  • Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
  • Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
  • If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
  • LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs

Low compensation

  • The LIVE/DEAD™ Fixable Violet Stain was selected based on its fluorescent properties to minimize compensation between other violet dyes and dyes that excite off of the 488 nm blue laser
  • The violet-fluorescent reactive dye has an excitation maximum of ∼416 nm and is excited well with the 405 nm violet laser
  • It has an emission maxima of ∼451 nm, so it can be collected in the first channel on most violet laser flow cytometers
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Cell Type Eukaryotic Cells
Color Violet
Content And Storage Contains 5 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Description LIVE/DEAD™ Fixable Violet Dead Cell Stain Kit, for 405 nm excitation
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Stain
Dye Type LIVE/DEAD™ Fixable Violet Dead Cell Stain
Emission 451
Excitation Wavelength Range 416 nm
Form Solid
Format Tube(s)
Product Line LIVE/DEAD
Quantity 200 Assays
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.